Protease inhibitor cocktail, Plant ProteaseArrest™
Leverandør: G Bioscienes
786-332
733-1948EA 2625 NOK
733-1948
Protease inhibitor cocktail, Plant ProteaseArrest™
Cellekultursupplementer og tilsetninger
A broad range, 100X concentrated, ready-to-use protease inhibitor cocktail, Plant ProteaseArrest™ inhibits plant serine, cysteine and other plant specific proteases including aminopeptidases, aspartic and metalloproteases.
  • Plant specific protease inhibitor cocktail
  • 100X concentrated, ready-to-use solution
Applications inhibition of protease activity in protein preparations of plant lysates and protection of proteins from proteolysis in such applications as electrophoresis, purification, storage and assays.