Protease inhibitor cocktail, Recom ProteaseArrest™
Leverandør: G Bioscienes
786-376
733-1997EA 2500 NOK
733-1997
Protease inhibitor cocktail, Recom ProteaseArrest™
Cellekultursupplementer og tilsetninger
Recom ProteaseArrest™ is a broad range, bacterial, 100X concentrated, ready to use protease inhibitor cocktail. Recom ProteaseArrest™ offers greater protection for recombinant proteins expressed and purified from bacteria. Recom ProteaseArrest™ inhibits bacterial serine, cysteine, metallo- and other bacterial specific proteases, including aminopeptidases and aspartic proteases.