Magnetic separation devices, Axygen®
Leverandør: Corning
IMAG-12T IMAG-96P
733-2133EA 12525 NOK
733-2133 733-2134
Magnetic separation devices, Axygen®
Stativer for rør Magnetisk seperasjonsstativ
The magnetic separation devices (MSD) are essential in any paramagnetic bead-based purification process. These IMAG™ devices are designed for a manual process.
  • Single tube or 96-well format
  • Applications include nucleic acid purification, cell based assays and antobody and protein purifications
  • Strong magnets enable separation times of less than 30 seconds
  • Manual operation for either low or high throughput