qScript™ XLT One-Step RT-qPCR ToughMix®
Leverandør: Quantabio
95134-500 95133-100 95132-100 95132-500 95133-500 95134-100 95132-02K 95133-02K 95134-02K 95132-05K 95134-05K
733-2234EA 12050 NOK
733-2234 733-2162 733-2161 733-2230 733-2232 733-2163 733-2908 QUNT95133-02K QUNT95134-02K 733-2909 QUNT95134-05K
qScript™ XLT One-Step RT-qPCR ToughMix®
Nukleinsyre reagenser qPCR/RT-qPCR Enzymes and Kits
qScript™ XLT One-Step RT-qPCR ToughMix® is a ready to use, highly sensitive master mix for reverse transcription quantitative PCR (RT-qPCR) of RNA templates.
  • Overcomes common inhibitors including polysaccharides, heme/haemoglobin, humic acid and melanin
  • Suitable for fast or standard qPCR cycling conditions
  • Broad dynamic range ensures reliable data from precious samples
  • Multiplexing enabled - supports detection of up to four targets
qScript™ XLT One-Step RT-qPCR ToughMix® is a versatile and robust RT-qPCR reagent that provides maximum sensitivity and PCR efficiency with a variety of fluorogenic probe chemistries, including TaqMan® hydrolysis probes.