PerfeCTa® qPCR ToughMix®
Leverandør: Quantabio
95112-012 95112-05K 95112-250 95114-012 95114-05K 95114-250 95113-012 95113-05K 95138-012 95138-05K 95139-012 95139-05K 95140-012 95140-05K
733-2090EA 26475 NOK
733-2090 733-2091 733-2092 733-2096 733-2097 733-2098 733-2093 733-2094 733-2243 733-2244 733-2247 733-2248 733-2251 733-2252
PerfeCTa® qPCR ToughMix®
Nukleinsyre reagenser qPCR/RT-qPCR Enzymes and Kits
PerfeCTa® qPCR ToughMix® is a 2X concentrated ready to use reaction cocktail for PCR amplification of DNA templates that overcomes many known inhibitors of PCR often present in crude samples extracted from environmental specimens, plant tissues or animal tissues. It is a versatile and robust Real-Time qPCR reagent that provides maximum sensitivity and PCR efficiency with a variety of fluorogenic probe chemistries, including TaqMan® hydrolysis probes. PerfeCTa® qPCR ToughMix® contains all required reaction components, except primers, probe(s) and DNA template. The light blue colour of the AccuVue™ tracer dye simplifies reaction assembly in white, or clear plates and helps to minimise pipetting or mixing errors. It does not interfere with qPCR performance or affect the stability of the product.
A key component of PerfeCTa® qPCR ToughMix® is an ultra-pure, highly processive thermostable DNA polymerase that is combined with high avidity monoclonal antibodies. This proprietary polymerase mix is highly resistant to PCR inhibitors and provides an extremely stringent automatic hot start allowing reaction assembly, and temporary storage, at room temperature prior to PCR amplification. PerfeCTa® qPCR ToughMix® delivers exceptional performance with either fast or conventional PCR cycling protocols.
Leveringsinformasjon: Kits are available with or without ROX reference dye. All kits include 2X reaction buffer containing optimised concentrations of MgCl₂, dNTPs (dATP, dCTP, dGTP, dTTP), hot start DNA polymerase, AccuVue™ blue qPCR dye, and stabilisers. UNG versions include uracil DNA glycosylase.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Applikasjon qPCR probe
Inhibitorrik prøve Ja