UDI adapters (1-96), sparQ

Leverandør: Quantabio

95211-096
733-2897EA 17200 NOK
733-2897
UDI adapters (1-96), sparQ
Nukleinsyre reagenser Nukleinsyrerensingsreagenser og -kit

sparQ UDI Adapters (1-96) are unique dual-indexed adapters designed to be used on illumina sequencing platforms. It is compatible with both DNA and RNA libraries. The use of UDI’s minimises index hopping events while also enabling greater multiplexing on flow cells.


  • Flexible pooling: Multiplex up to 96 samples per sequencing run
  • Improved performance: Prevents index hopping and enhances demultiplexing accuracy
  • Optimised: For maximum ligation efficiency with sparQ library prep kits
  • Multiple applications: Including whole genome (with amplification or PCR-free), target enrichment, whole transcriptome sequencing and many more

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Applikasjon
Target molecule
Refine target molecule
Sample type
Method / Format

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR