Laminin cell culture flasks, Corning® BioCoat®
Leverandør: Corning
354522
734-0181EA 9775 NOK
734-0181
Laminin cell culture flasks, Corning® BioCoat®
Kolber Celledyrkningsflasker
Laminin, a major structural component of basement membranes, is a 900 kD glycoprotein composed of three polypeptide chains with a multidomain structure. Laminin has many varied funcitons that are mediated by binding to various components of the basement membrane (for example, collagen IV and heparan sulphate proteoglycan) and to cell surface receptors. Corning® BioCoat® Laminin flasks, coated with mouse laminin, are manufactured in a highly controlled environment and rigorously tested to assure product consistency and performance.
Applications include promotion of cell attachment andMore Product Information