Collagen IV-coated flasks, Corning® BioCoat®
Leverandør: Corning
354523 354528
734-0182EA 9500 NOK
734-0182 734-0185
Collagen IV-coated flasks, Corning® BioCoat®
Kolber Celledyrkningsflasker
Collagen IV is a ubiquitous component of basement membranes. It plays a role in the regulation of cell growth, differentiation and adhesion, as well as tissue formation. Corning® BioCoat™ Collagen IV flasks are tissue culture vessels with a uniform application of mouse collagen type IV. These flasks are manufactured in a highly controlled environment and are rigorously tested to assure product consistency and performance.
Applications include promotion of cell attachment and spreading, cell differentiation and neurite outgrowth, increased proliferation of PC 12 cells, studies of effectsMore Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact
Miljøsertifikat ISO 14001 - Bedford