Dialyseslange, Spectra/Por® 7
Leverandør: Spectrum Laboratories
128358 132103 132104 132105 132107 132108 132109 132110 132111 132112 132113 132114 132115 132116 132117 132118 132119 132120 132121 132122 132123 132124 132125 132126 132127 132128 132129 132130 132131 132120T 132112T 132103T 132107T 132111T 132119T
734-3502EA 8575 NOK
734-3502 734-0605 734-0606 734-0607 734-0608 734-0609 734-0610 734-0611 734-0612 734-0613 734-0614 734-0615 734-0616 734-0617 734-0618 734-0619 734-0620 734-0621 734-0622 734-0623 734-0624 734-0625 734-0626 734-0627 734-0628 734-0629 734-0630 734-0631 734-0632 514-0913 514-0908 514-0904 514-0905 514-0907 514-0912
Dialyseslange, Spectra/Por® 7
Slanger Dialyseslanger
MWCO 1 to 50 kD, regenerated cellulose.
  • Transparent and flexible
  • Minimum sulphur and heavy metal contamination
Chemically treated to remove any trace levels of heavy metals and sulfide compounds that may be present in dry Spectra/Por® 1 to 5 and pre-wetted Spectra/Por® 6 Standard RC membranes.
Leveringsinformasjon: Supplied wet in 0,1% sodium azide.