Cellekulturinnsatser med fibronektin, Corning® BioCoat™ FluoroBlok™
Leverandør: Corning
354597
734-1053EA 13400 NOK
734-1053
Cellekulturinnsatser med fibronektin, Corning® BioCoat™ FluoroBlok™
Mikroplater Innsatsplater
Corning® BioCoat™ FluoroBlok™ fibronectin-coated cell culture inserts packaged ready to use in 24-well Falcon® companion plates.
  • Convenient, individual inserts packaged in a one-piece, automation friendly 24-multiwell format
  • Combined benefits of Falcon® inserts coated with HFN for enhanced cell attachment and growth
  • Individual inserts contain a 3,0 µm pore size clear PET membrane
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact
Miljøsertifikat ISO 14001 - Bedford