Kammerobjektglass, Corning® BioCoat™ poly-D-lysin/laminin
Leverandør: Corning
354688
734-1072EA 10300 NOK
734-1072
Kammerobjektglass, Corning® BioCoat™ poly-D-lysin/laminin
Objektglass Objektglass med kammer
These culture slides have a uniform application of poly-D-lysine/laminin.
  • Tested to promote neurite outgrowth
  • NG108 rat glioma/mouse neuroblastoma cells
  • PS vessel lid together with removal tool