Utbyggingssett for rollerapparat, Wheaton®
Leverandør: DWK Life Sciences
W348930-CH
734-1170EA 8050 NOK
734-1170
Utbyggingssett for rollerapparat, Wheaton®
Rollerapparater
Attaches quickly and easily to the small bottle bench top roller culture system. Holds an additional two rollers per kit. These units will accept two additional decks. Kit consists of assembled roller deck, four support posts, drive belt, necessary hardware and assembly instructions.
Vekt: 2,3 kg