VWR®, Drosophila Vials
Leverandør: VWR Collection
734-1255EA 2200 NOK
734-1255
VWR®, Drosophila Vials
Vialer
These PS vials have a flat bottom straight wall design for convenient racking, stacking and filling.
The tray packed 10×10 row configuration is great for quick, effortless filling.
Bulk packed vials are layered with open-end orientation for convenient handling and smaller case size.
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904