Hollow fibre Implant membranes, KrosFlo®
Leverandør: Spectrum Laboratories
S9320101 M138615
734-1387EA 3050 NOK
734-1387 734-1384
Hollow fibre Implant membranes, KrosFlo®
Slanger Dialyseslanger
KrosFlo® hollow fibre membranes for implant technology provide a true-to-life response for drug screening and cancer research. Perfectly suited for implantation, the modified Polyvinylidene Difluoride (mPVDF) hollow fibre membrane is biocompatible, resistant to cell adhesion, hydrophobic and resistant to a wide variety of organic solvents, including most aqueous acids and bases. Furthermore, these hollow fibres can be heat sealed and autoclaved without affecting the molecular weight cut off (500 kDa MWCO) or changing the membrane performance.
  • Isolate immune systems cells, viruses and