24 clear not treated reservoir, sterile, HTS Transwell®
Leverandør: Corning
3395
734-1558EA 3950 NOK
734-1558
24 clear not treated reservoir, sterile, HTS Transwell®
Mikroplater HTS-mikroplater
For use as reservoir tray with HTS Transwell permeable supports.
  • Not treated for cell attachment
  • Same footprint as standard 24 well plate but with single rectangular well