Aspirating pipettes, Costar®
Leverandør: Corning
9186 9099 4975 9016
734-1779EA 6950 NOK
734-1779 734-1778 612-5122 612-3720
Aspirating pipettes, Costar®
Pipetter Aspireringspipetter
PS pipettes, non graduated, unplugged, sterile.
  • One piece PS construction
  • For aseptically aspirating liquid by using vacuum suction
  • Certified non pyrogenic
Forpakning: Bulk (1 ml) or individually wrapped (2 and 5 ml).