Aspirating pipette, Corning®
Leverandør: Corning
4930
734-1997EA 5675 NOK
734-1997
Aspirating pipette, Corning®
Pipetter Aspireringspipetter
This aspirating device is used for the removal of liquids from a variety of lab vessels. It is designed for use with an in-house vacuum system. The aspirator allows precise fingertip control of aspiration and presents a sensible alternative to the use of glass pasteur pipettes for supernatant disposal.
  • Designed for a comfortable fit in either hand
  • Can be used with a variety of collection devices
Designed for use with a vacuum pump or an in-house vacuum system and a suitable collection vessel
Leveringsinformasjon: Supplied with handpiece, grommetMore Product Information