Accessories for aspirating pipettes, Costar®
Leverandør: Corning
4931
734-1998EA 3200 NOK
734-1998
Accessories for aspirating pipettes, Costar®
Pipetter Aspireringspipetter
This is an 8-channel adapter that fits on the Costar® aspirator so that it can be used to remove media and solutions from 8 wells simultaneously on a 96 well plate.