Caps for cell culture flasks, EasYFlasks™, Nunclon™Δ
Leverandør: Thermo Fisher Scientific
133001
734-2144EA 1300 NOK
734-2144
Caps for cell culture flasks, EasYFlasks™, Nunclon™Δ
Lokk Flaskelokk
Ideal for cell culture flasks. Offered in two different types vent/close and filter.
  • Sterile
  • HDPE closure