Caps for cell culture flasks, Nunc™ Cell Factory™
Leverandør: Thermo Fisher Scientific
147074
734-2152EA 1450 NOK
734-2152
Caps for cell culture flasks, Nunc™ Cell Factory™
Lokk Lokk til reagensglass
These caps reduce the need for open interventions and support the use of aseptic methods for fluid handing relating to filling, inoculation, feeding and harvesting of the cell factory system.
Further the development of applications requires a more closed cell culture system using Thermo Scientific™ Nunc™ Cell Factory™ accessories.