Carboxyl cell culture flasks, Corning® PureCoat™
Leverandør: Corning
354778 356780
734-2213EA 695 NOK
734-2213 734-2408
Carboxyl cell culture flasks, Corning® PureCoat™
Kolber Celledyrkningsflasker
PS, with blue screw cap.
Corning® PureCoat™ carboxyl flasks provide an enhanced tissue culture surface. The negatively charged surface, manufactured using a proprietary thin-film coating technology, offers improved attachment, increased proliferation, and enhanced recovery from freeze-thaw for a broad range of primary, transfected and fastidious cells that exhibit poor attachment properties on standard tissue culture (TC) surfaces in serum-free/serum-reduced conditions.
NB: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, whereMore Product Information