ECM mimetic cultureware plates, Corning® PureCoat™
Leverandør: Corning
356270 356271
734-2494EA 6450 NOK
734-2494 734-2495
ECM mimetic cultureware plates, Corning® PureCoat™
Mikroplater Celledyrkningsplater
Corning ECM mimetic surfaces and substrates contain biologically active, synthetic, animal-free peptides that have been rationally designed to mimic the cell attachment motifs of native ECM proteins. The peptides enable optimal cell binding and signaling in a broad range of serum-free, xeno-free, and animal-free media formulations, supporting the propagation and differentiation of a range of stem, progenitor and primary cell types.
  • Available peptides: Human collagen I and human fibronectin
  • Delivers similar adhesion capabilities as the native protein,