Microdialysis devices, ready-to-use, Micro Float-A-Lyzer®
Leverandør: Spectrum Laboratories
F235055 F235065 F235049 F235051 F235053 F235070 F235061 F235057 F235067 F235069 F235058 F235059 F235071 F235063
734-2589EA 4075 NOK
734-2589 734-2595 734-2586 734-2587 734-2588 734-2598 734-2593 734-2590 734-2596 734-2597 734-2591 734-2592 734-2599 734-2594
Microdialysis devices, ready-to-use, Micro Float-A-Lyzer®
Slanger Dialyseslanger
Designed to maximise convenience and efficiency, the ready-to-use Micro Float-A-Lyzer® is ideal for the dialysis of very small sample volumes. Available in two volume sizes, 100 to 200 μl and 400 to 500 μl, the Micro Float-A-Lyzer® features a proprietary biotech grade cellulose ester (CE) membrane incorporated into a pre-assembled, leakproof microdialysis device. Available in 7 concise MWCO’s with colour-coded caps, CE is a synthetic, low-protein binding membrane with no heavy metal and sulfide contaminants. The pre-formed tubular geometry also limits volume increase and sample dilution. TheMore Product Information