VWR® Cotton Balls
Leverandør: VWR Collection
734-2671EA 3450 NOK
734-2671
VWR® Cotton Balls
Bomull og glassull
Thorough machine processing provides consistent uniformity of shape and size for maximum efficacy in vial plugging. These high quality balls are very effective at controlling mite infestation.
  • Fits narrow vials
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904