Automated cell counter, EVE™
Leverandør: NANOENTEK
EVE-MC-EU
734-2675EA 41075 NOK
734-2675
Automated cell counter, EVE™
Celletellere Elektroniske celletellere
EVE™ is a benchtop automatic cell counter designed to measure cell count and viability accurately and precisely. It uses the standard trypan blue technique.
  • Fine distinction of clumped cells: Accurate results with advanced analysis algorithm
  • Broad range of cell sizes and types: Primary (tissue and blood) cell lines and stem cells
  • User friendly: Touch screen LCD without any maintenance needed
  • Data store analysis: 500 test results, transfer to PC via USB drive
  • Cell size gating: Select range of cell size using gating function