VWR®, Cellekulturskåler
Leverandør: VWR Collection
734-2319EA 2850 NOK
734-2319 734-2318 734-2320 734-2321 734-2322 734-2317 734-2754 734-2793 734-2794 734-2795 734-2796 734-2797 734-2814 734-2815 734-2816 734-2817 734-2818
VWR®, Cellekulturskåler
Dyrkningsplater Cellekulturskåler
VWR® cell and tissue culture dishes are available with a choice of surface treatments.
  • Flat bottom and uniform wall thickness ensures distortion-free viewing
  • Optically clear, making them suitable for microscopy
  • Dish surface is smooth and free from striations to maximise usable growth area
  • Rim on the bottom side of the lid (except 35 or 150 mm dishes) mates with the dish brim for easy and secure stacking
  • Sterilised by electron beam irradiation, DNase- and RNase-free and non pyrogenic
Increased cell attachment surface: This highly hydrophilic surfaceMore Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact
Miljøsertifikat ISO 14001/Certificate No.: 04923E00243R0M