384-well microplates, ShallowWell, standard height
Leverandør: Thermo Fisher Scientific
264704
734-2876EA 10300 NOK
734-2876
384-well microplates, ShallowWell, standard height
Mikroplater HTS-mikroplater
These standard height plates are suitable for automation, without the need to make adjustments to robotics, plate readers or liquid handlers.
  • Untreated hydrophobic polystyrene suited for growth of suspension cultures
  • ANSI/SBS standard footprint