Mikroplater, standardhøyde, 384 ShallowWell, Nunc™
Leverandør: Thermo Fisher Scientific
264705
734-2877EA 4275 NOK
734-2877
Mikroplater, standardhøyde, 384 ShallowWell, Nunc™
Mikroplater HTS-mikroplater
Nunc™ 384 ShallowWell standard height, black and white plates save costs in samples and reagents by reducing the total well volume to 25 μl. The standard height of the plate allows the use of automation without making adjustments to robotics, readers or liquid handlers.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact
Miljøsertifikat NAR Certificate of Retirement