Cell analysis slides, Arthur™
Leverandør: NANOENTEK
AC0050
734-2896EA 2575 NOK
734-2896
Cell analysis slides, Arthur™
Objektglass Objektglass med kammer
Arthur™ Cell analysis slides are designed for use with the Arthur™.
  • Assay for cells including GFP&RFP expression, apoptosis, cell viability, cell cycle, and cell counting
  • Eliminates washing steps and cross contamination between samples (fully disposable)
Size: 110×24×1,9 mm