VWR® Single-Well Cell Culture Plates
Leverandør: VWR Collection
734-2966EA 4300 NOK
734-2966 734-2977
VWR® Single-Well Cell Culture Plates
Mikroplater Celledyrkningsplater
Single-well cell culture plates, made from clear PS, are available plasma-treated for consistent cell attachment and growth, or non-treated for use in applications where cell attachment is not desired.
  • Increased internal plate dimension, providing larger area for cell growth
  • Standard SBS footprint makes them compatible with standard equipment and automated systems
  • Free from DNase, RNase, and human DNA, certified non-pyrogenic, non-cytotoxic
  • Sterilised by electron beam irradiation
TC-treated: These dishes are vacuum-gas plasma or corona treated forMore Product Information