VWR®, Cell Lifters
Leverandør: VWR Collection
734-2978EA 3275 NOK
734-2978 734-2979
VWR®, Cell Lifters
Celleskrapere
Flat blade cell lifter for manual harvesting of cells, manufactured from high grade PE. Designed for removing cells from flasks or dishes.
  • Bevelled edge blade on one end and narrow blade or J-hook on the other
  • Sterilised by electron beam irradiation
  • Free from DNase, RNase and non-pyrogenic