OmniTop Sample Tubes®-sammenstillinger, forhåndssterilisert engangsbruk
Leverandør: Avantor Fluid Handling
734-2986EA 16550 NOK
734-2986 734-3360 734-2993 734-2994 734-2995 734-2985 734-2987 734-2990 734-2989 734-2991 PAWB15CTAPPCFDSL00 PAWB50CTA-CF-DSL-S
OmniTop Sample Tubes®-sammenstillinger, forhåndssterilisert engangsbruk
Rør Reagensrør
OmniTop Sample Tubes® are a convenient device that can be used to obtain fluid samples. Each OmniTop tube comes with a pre-attached 0,2 µm vent filter and 18" of tubing (C-Flex®, TYGON®, silicone or PharMed®).
  • Customisable cap system
  • Unique cap design allows for complete customisation
  • Ability to use different ID and OD tubing Ø
  • Wide selection of tubing materials
  • Easily configured with virtually any type of tubing
  • Single-use
  • Cost effective
  • Reduced assembly and installation time
  • Eliminate cleaning validation
  • Flexible manufacturing
  • Design allows quick