Kulturflaske, dobbel sidearm, Celstir®, WHEATON®
Leverandør: DWK Life Sciences
356873 356875 356876 356879 356884 356889 356882 356887 356890
734-3006EA 9125 NOK
734-3006 734-3007 734-3008 734-3009 734-3011 734-3013 734-3010 734-3012 734-3014
Kulturflaske, dobbel sidearm, Celstir®, WHEATON®
Kolber Spinnerflaske
Flaske i borsilikatglass, røreenhet av PTFE og glass, lokk med PTFE og silikonpakning.
  • Adjustable paddle blade impeller allows better mixing
  • Impeller does not protrude through the top cap, thereby maximising incubator space and reducing the risk of contamination
  • Addition of the bottom dimple to flasks 125 ml and larger improves circulation and reduces the accumulation of cells in the centre of the flask
  • Flasks 500 ml or larger have a 45 mm side arm to be used as an air vent, media inlet or outlet, innoculation port, pH probe inlet, or other application