Cell culture inserts, 2 in 1, cellGrade™ plus
Leverandør: Brand
782700 782740 782720 782701 782706 782741 782707 782721 782710 782746 782726 782711 782716 782727 782747 782717 782750 782730 782731 782751 782736 782756 782737 782757
734-3267EA 7350 NOK
734-3267 734-3283 734-3275 734-3268 734-3269 734-3284 734-3270 734-3276 734-3271 734-3285 734-3277 734-3272 734-3273 734-3278 734-3286 734-3274 734-3287 734-3279 734-3280 734-3288 734-3281 734-3289 734-3282 734-3290
Cell culture inserts, 2 in 1, cellGrade™ plus
Mikroplater Innsatsplater
Cell culture inserts with cellGrade™ plus treated membranes for improved cell attachment and growth.
  • Can be used as a hanging insert (fits nearly all multiwell plates) or, for bigger wells and cuture dishes, as a standing insert
  • cellGrade™ plus surface for excellent cell attachment and growth
  • Highly transparent membranes give good visibility of cell cultures
  • High pore density contributes to good permeability
  • Innovative packaging provides for efficient unpacking and reduces waste
Bestillingsinformasjon: Inserts are available for 6-, 12- and 24-wellMore Product Information