Nunc™ Microwell 96-Well Microplate with Nunclon™ Supra Surface
Leverandør: Thermo Fisher Scientific
167013
734-3635EA 79 NOK
734-3635
Nunc™ Microwell 96-Well Microplate with Nunclon™ Supra Surface
Mikroplater Celledyrkningsplater
Experience optimal performance with Nunc™ plates with Nunclon™ Supra surface treatment for culturing a wide variety of fastidious adherent cells. Intended for general cell culture applications, the Nunclon™ Supra surface is more hydrophilic than the Nunclon™ Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
  • Consistent lot-to-lot performance
  • Sterile with SAL 10⁻⁶
  • Non-cytotoxic
  • Non-pyrogenic
  • Biocompatible per USP Class VI for raw materials
  • Five years shelf life
The Nunclon™ Supra surface supports serum-More Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact
Miljøsertifikat IRECS