Corning® 10-Layer CellCube® Module, Corning
Leverandør: Corning
3200
734-4051EA 14500 NOK
734-4051
Corning® 10-Layer CellCube® Module, Corning
Bioreaktorer
The CellCube Module is an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment.
It is comprised of a series of parallel, styrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates. Each culture plate receives proprietary tissue culture treatment prior to assembly to ensure dense, uniform cell growth. These CellCube Modules provide: a series of parallel, polystyrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates, 8,500 cm² polystyrene growth surface optimally treated for theMore Product Information