J.T.Baker®, Multipurpose Alkaline Detergent, RBS 35 concentrate
Leverandør: Avantor
7348.5000EA 1300 NOK
7348.5000
J.T.Baker®, Multipurpose Alkaline Detergent, RBS 35 concentrate
Rengjøringsløsninger Detergenter og overflatevasking
RBS 35 concentrate is used for manual cleaning, soaking and ultrasonic baths. These surfactants are alkaline, foaming and insensitive to water hardness.
  • High wetting and emulsifying power
  • No risk of calcareous deposits when diluted in tap water
  • Total rinse
Suitable for cleaning glassware, laboratory equipments and for cleaning parts made of glass, stainless steel, injection moulds and production equipment.
Typical concentration for use: 2%