Micro-volume spectrophotometer, SpectraMax® QuickDrop
Leverandør: MOLECULAR DEVICES
QUICKDROP
735-0350EA 0 NOK
735-0350
Micro-volume spectrophotometer, SpectraMax® QuickDrop
Spektrofotometre og fotometre Biofotometere
An affordable, one touch, full spectrum micro volume spectrophotometer, ideal for quantification of very small quantities of DNA, RNA, proteins, amino acids and peptides – a critical first step in many molecular biology, biochemistry and genetics workflows. Additionally, this unit is also suitable for many more general absorbance-based applications, including quality control and testing.
  • Intuitive touch screen interface with pre-configured analysis methods simplifies data acquisition and analysis whilst still allowing the creation of customised experiments