Imaging cytometer, SpectraMax® MiniMax™ 300
Leverandør: MOLECULAR DEVICES
MINIMAX300
735-0380EA 0 NOK
735-0380
Imaging cytometer, SpectraMax® MiniMax™ 300
Mikroplatelesere
Designed specifically for the SpectraMax® i3 multi-mode detection platform, the field-upgradeable SpectraMax® MiniMax™ 300 imaging cytometer enables quick inspection of cells and provides insight into phenotypic changes that are important for understanding cytotoxicity, cell proliferation, and protein expression. Designed to enable cellular imaging capabilities, this cytometer unit now features StainFree™ cell counting technology, additional Brightfield analysis features, and fluorescent green and red detection channels.
  • Simplified workflow: Add cellular imaging assays in a small