DispenCell™ Single-Cell Dispenser
Leverandør: MOLECULAR DEVICES
DISPENCELL
735-0641EA 0 NOK
735-0641
DispenCell™ Single-Cell Dispenser
Koloniplukkere
The DispenCell™ Single-Cell Dispenser is an automated laboratory instrument designed for fast, easy and gentle single-cell isolation. DispenCell integrates seamlessly into your laboratory workflow, with a plug-and-play approach. Flexible and effortless, DispenCell operates equally under sterile conditions in a culture hood, or on a simple benchtop.
  • The small footprint of the DispenCell™ allows it to fit perfectly into any laboratory setting: Under a hood to work under sterile conditions, on a bench top for routine seeding, or in a pre-existing automated