Robotic tips for Beckman®
Leverandør: Thermo Fisher Scientific
915-021 917-021 918-262 918-261 918-021 919-262 919-261 919-021 919-021G 917-262 935-262 3762 935-261 935-021 915-262 912-021 912-261 912-262 915-261 917-261 918-011 918-021-05 918-251 918-252 918-261-05 918-262-05 919-011 919-021-05 919-251 919-252 919-261-05 919-262-05 2790 919-261G 919-262G 2760 916-021 916-261 916-262
736-0015EA 3550 NOK
736-0015 736-0016 736-0090 736-0020 736-0019 736-0099 736-0096 736-0021 736-0094 736-0083 737-2014 736-0038 737-2012 737-2011 736-0077 736-0013 736-0070 736-0071 736-0075 736-0017 736-0018 736-0086 736-0087 736-0088 736-0089 736-0091 736-0092 736-0093 736-0022 736-0095 736-0097 736-0100 736-0001 736-0098 736-0101 736-0000 736-0616 736-0617 736-0618
Robotic tips for Beckman®
Pipettespisser
High quality tips, tested on the workstations for which they were created, to ensure compatibility and a secure fit. Certified free of RNase, DNase, DNA, RNA, pyrogens and ATP.
  • Transparent PP
  • Tips with ART® filters: Eliminate any risk of cross-contamination between samples; designed for applications with DNA, RNA as well as radioactive and infectious samples
  • Low retention tips: Hydrophobic tips retain less sample which decreases the amount of reagent loss while significantly increasing result reproducibility
  • Sterile tips: Sterilised by gamma radiation