Adhesive seal, AMPLIseal™
Leverandør: Greiner Bio-One
676040_100
736-0354EA 2275 NOK
736-0354
Adhesive seal, AMPLIseal™
Forseglinger Filmer og folier til mikroplateforsegling
AMPLIseal™ is a self-adhesive sealing film notable for its low autofluorescence, in particular in the wavelength range critical for Real Time PCR. It consists of a thick PP film coated with a transparent acrylate adhesive. The adhesive layer is protected by a peelable polyester film. The strongly adhering acrylate adhesive provides reliable sealing, thereby minimising evaporation but without influencing the PCR reaction or the fluorescence measurement.
  • Highly transparent with minimal autofluorescence
  • Ideal for Real Time PCR