VWR®, Robotspiss, for Beckman® Biomek FX/NX
Leverandør: VWR Collection
736-0611EA 1225 NOK
736-0611
VWR®, Robotspiss, for Beckman® Biomek FX/NX
Pipettespisser
Gjennomsiktig PP, kan autoklaveres.
  • RNase/DNase, DNA and endotoxin-free
  • Sustainable design based on virgin resins
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact,Sustainable Packaging