Nucleic acid and protein purification systems, KingFisher™ Flex
Leverandør: Thermo Fisher Scientific
5400610 5400640 5400620 5400630
731-0139EA 0 NOK
731-0139 736-0684 731-0140 736-0683
Nucleic acid and protein purification systems, KingFisher™ Flex
Renseaparat for nuklein syrer
The KingFisher™ magnetic particle processors are designed to automate time consuming sample processing of nucleic acids, proteins or cells from virtually any source (blood, cell cultures, tissue lysates, soil, faeces, for example). The KingFisher™ family consists of instruments with different throughput and volume capacities to fulfil individual sample processing needs. Each system consists of an instrument, specially designed plastics, and software. The KingFisher™ systems offer rapid and reproducible purification of high quality DNA, RNA, proteins and cells for various types of downstreamMore Product Information
Bestill nå
SPESIFIKASJONER
Modell KingFisher Flex with 96 PCR head KingFisher Flex med 24 dype brønnhoder KingFisher Flex with 96 KF head KingFisher Flex med 96 dype brønnhoder
Prøvevolum 20 - 100 µl 200 - 5000 µl 20 - 100 µl 50 - 1000 µl
Environmentally Preferable
Kapasitet (prøver/gang) 96 24 96 96
Effektivitet ved innsamling av partikler >95% >95% >95% >95%
Størrelse på magnetiske partikler ~ >1 μm ~ >1 μm ~ >1 μm ~ >1 μm
Platetyper 20 - 100 μl 200 - 5000 μl 20 - 250 μl 50 - 1000 μl
Kamspisser (engangsutstyr) Special design, 96 format Special design, 96 format
Inkubering Heating block temperature from RT to +115 °C Heating block temperature from RT to +115 °C
Innebygd programvare Space for ~100 internal protocols Space for ~100 internal protocols
Grensesnitt USB or serial RS232C port USB or serial RS232C port
Tastatur/display START/STOP/PAUSE/OK, four arrow keys, two keys to rotate the turntable, colour LCD START/STOP/PAUSE/OK, four arrow keys, two keys to rotate the turntable, colour LCD
Robotkompatibilitet Yes Yes Yes Yes
Vekt 28 kg 28 kg 28 kg 28 kg
B×D×H 680×600×380 mm 680×600×380 mm 680×600×380 mm 680×600×380 mm