Natriumacetat trihydrat 99.0-101.0%, granular, AR® ACS, Macron Fine Chemicals™

Leverandør: Avantor Performance Materials
Warning

7364-04EA 0 NOK
7364-04 7364-20 7364-88
Natriumacetat trihydrat 99.0-101.0%, granular, AR® ACS, Macron Fine Chemicals™
Natriumacetat trihydrat

AR—The standard Macron Fine Chemicals™ grade of analytical reagents; suitable for laboratory and general use. If the reagent also meets the requirements of the American Chemical Society Committee on Analytical Reagents, it will be denoted as an AR (ACS) reagent. 

Formel: H₃CCOONa·3H₂O
Kokepunkt: 120…123 °C (1013 hPa)
Smeltepunkt: 58 °C
Tetthet: 1,4 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00071557
CAS-nummer: 6131-90-4
EINECS: 204-823-8
Merck Indeks: 13,08642
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use 99.0 - 101.0 %
ACS - Assay (CH₃COONa·3H₂O) 99.0 - 101.0 %
ACS - Calcium (Ca) ≤0.005 %
ACS - Chloride (Cl) ≤0.001 %
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Insoluble Matter ≤0.005 %
ACS - Iron (Fe) ≤5 ppm
ACS - Magnesium (Mg) ≤0.002 %
ACS - Phosphate (PO₄) ≤5 ppm
ACS - Potassium (K) ≤0.005 %
ACS - Substances Reducing Permanganate Passes Test
ACS - Sulfate (SO₄) ≤0.002 %
ACS - pH of 5% Solution at 25°C 7.5 - 9.2
Appearance (fine, colorless granules)
Product Information (not specifications):


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR