Polypropylene microplates, 96- and 384-well
Leverandør: EPPENDORF
0030627.300
737-0237EA 20800 NOK
737-0237
Polypropylene microplates, 96- and 384-well
Mikroplater HTS-mikroplater
These polypropylene microplates feature high transparency and easy to read alphanumeric labelling, making the identification of the correct plate or an individual well quick, easy, and less error-prone. Black and white versions are available for fluorescence or luminescence detection, offering excellent signal-to-noise ratios for improved sensitivity.
  • Highest transparency for better sample visibility
  • OptiTrack® matrix for easy and fast well identification
  • g-safe® for time saving centrifugation up to 6000×g
  • Well designed for maximum sample recovery with
More Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact
Miljøsertifikat Renewable Energy Certificate, ISO 14001