Benzosyre, Supelco®

Leverandør: Merck
Warning


M-Clarity™ quality level = MQ100

73983-5G-F
73983-5G-FEA 565 NOK
73983-5G-F
Benzosyre, Supelco®
Benzosyre
Formel: C₇H₆O₂
MW: 122.12 g/mol
Kokepunkt: 249
Smeltepunkt: 121…123 °C
Tetthet: 1.316 g/cm³ (20 °C)
Flash punkt: 121 °C (closed cup)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002398
CAS-nummer: 65-85-0
EINECS: 200-618-2
REACH: 01-2119455536-33
Merck Indeks: 13,01092
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=544a7c1a30a1e5f8cc337a2d15434cc6e6913a81&vid=e3186376027deeae82999b009919941a36bb3e5b&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR