Supelco® TraceCERT® Organic Reference Standard, Tryptamine hydrochloride
Leverandør: Merck
74099-50MG
74099-50MGEA 2625 NOK
74099-50MG
Supelco® TraceCERT® Organic Reference Standard, Tryptamine hydrochloride
Standarder Organiske standarder
Sertifiseringer: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
NB: P261 - P264 - P280 - P301 + P312 - P302 + P352 - P305 + P351 + P338