1-Nonanesulphonic acid sodium salt, Supelco®

Leverandør: Merck

Synonymer: Sodium 1-nonanesulphonate, Sodium nonane-1-sulphonate

MQ100

74316-10G-F
74316-10G-FEA 6420 NOK
74316-10G-F
1-Nonanesulphonic acid sodium salt, Supelco®
1-Nonanesulphonic acid sodium salt
Formel: C₉H₁₉NaO₃S
MW: 230.30 g/mol
MDL nummer: MFCD00025010
CAS-nummer: 35192-74-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a6ee6c5bf302e89ebf7705d4d943e60130361ea&vid=5d4bde5f6807da85672e9b18a35b36645049456b&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR