Natriumhydrogensulfat monohydrat, white crystals, AR®, Macron Fine Chemicals™

Leverandør: Avantor Performance Materials
Danger

7432-20EA 0 NOK
7432-20
Natriumhydrogensulfat monohydrat, white crystals, AR®, Macron Fine Chemicals™
Natriumhydrogensulfat monohydrat
Formel: NaHSO₄·H₂O
Tetthet: 2,1 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149210
CAS-nummer: 10034-88-5
EINECS: 231-665-7
FN: 3260
ADR: 8,III
Merck Indeks: 13,08658

Bestill nå

Spesifikasjon Testresultater

For Laboratory,Research,or Manufacturing Use
Solubility (10 in 50) Passes Test
Chloride (Cl) ≤0.001 %
Acidity (as H₂SO₄) 35 - 37 %
Arsenic (As) ≤0.0001 %
Insoluble Matter and NH₄OH Precipitate ≤0.005 %
Calcium and Magnesium Precipitate ≤0.005 %
Heavy Metals (as Pb) ≤0.0005 %
Trace Impurities - Iron (Fe) ≤0.001 %
Phosphate (PO₄) ≤0.001 %
Appearance (white crystals)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR